Learn More
Medchemexpress LLC Bet-in-19 (compound 146) | 1643947-30-1 | 99.7% | 333.39 g/mol | C19H19N5O | 10MM 1ML

Supplier: Medchemexpress LLC HY12523610MM 1ML
BET-IN-19 is a small-molecule inhibitor of BET family bromodomains used in epigenetic and oncology research. It inhibits tetra-acetylated histone H4 binding to BRD4 bromodomain 1 and has demonstrated cellular activity in reported assays (IC50 ≤0.3 μM). Supplied as a powder for laboratory research.
- Small-molecule BET bromodomain inhibitor for epigenetic and oncology research.
- Demonstrated cellular activity with IC50 ≤0.3 μM in reported assays.
- High purity (99.74%) suitable for in vitro studies.
- Molecular formula C19H19N5O, molecular weight 333.39 g/mol.
- Supplied as a powder; storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
- Soluble in DMSO at 100 mg/mL (≈299.95 mM); ultrasonic assistance may be required for dissolution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.