Learn More
Medchemexpress LLC Beta-1 | 2924598-24-1 | C37H28IN3S2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15207310MM/1ML
Encompass_Preferred
BETA-1 is the first twisted intramolecular charge transfer (TICT)-aggregation-induced emission (AIE) integration molecule. It emits cyan fluorescence in lipid droplets (LDs) and red fluorescence in mitochondria, enabling simultaneous and dual-color imaging of LDs and mitochondria both in vivo and in vitro.
  • Used for imaging of lipid droplets and mitochondria in live cells.
  • Enables simultaneous and dual-color imaging of lipid droplets and mitochondria.
  • Emits cyan fluorescence in lipid droplets and red fluorescence in mitochondria.
  • Applicable in vivo and in vitro studies.
Catalog No. 50-003-73299