Learn More
Medchemexpress LLC Beta 2 Microglobulin Antibody (YA935) | 10 UL

Supplier: Medchemexpress LLC HYP81260A10UL
Beta 2 Microglobulin Antibody (YA934) is a Mouse-derived and non-conjugated IgG2b monoclonal antibody, targeting beta 2 Microglobulin. It is for research use only.
- Host: Mouse
- Clonality: Monoclonal
- Species reactivity: Human
- Immunogen: Purified recombinant human β2-microglobulin protein
- Application: ELISA (1:10000 dilution)
- Sensitivity: Endogenous
- Conjugation: Non-conjugated
- Modification: Unmodified
- Isotype: IgG2b
- Appearance: Liquid
- Formulation: 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide, 0.05% BSA
- Function: Component of MHC, involved in peptide antigen presentation
- Subcellular localization: Secreted, cell surface
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.