Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-Acetoxyisovalerylshikonin | 69091-17-4 | 10 MM 1 ML
SDP

Supplier:  Medchemexpress LLC HYN218810MM/1ML

Encompass_Preferred

Beta-Acetoxyisovalerylshikonin is a naphthoquinone derivative isolated from *Arnebia euchroma*. It presents as a solid, typically brown to black, with a molecular formula of C23H26O8 and a molecular weight of 430.45. It is suitable for laboratory use and substance manufacturing.

  • Molecular weight of 430.45
  • Molecular formula C23H26O8
  • Solid appearance (brown to black)
  • Purity of 99.57%
  • Recommended storage: 4°C (solid), protected from light
  • Solvent storage: -80°C for 6 months, or -20°C for 1 month, protected from light
  • Soluble in DMSO (50 mg/mL, 116.16 mM)

Catalog No. 50-004-20627


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.