Learn More
AnaSpec Beta - Amyloid (1 - 42), HiLyteᵀᴹ Fluor 555 - labeled, Human, 0.1mg
Supplier:  AnaSpec AS6048001
Encompass
Sequence: HiLyte™ Fluor 555-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA, Molecular Weight 5366.57, This is a fluorescent (HiLyte™ Fluor 555)-labeled ß-Amyloid peptide, Abs/Em=551/567 nm.
Catalog No. NC0086764