Learn More
CPC Scientific H-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD013A
Encompass
ONE-LETTER SEQUENCE: LVFFAEDVGSNKGAIIGLMVGGVVMOLECULAR FORMULA: C110H178N26O31S1MOLECULAR WEIGHT:2392.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [156790-69-1], SYNONYMS: Amyloid β-Protein (17-40)RESEARCH AREA: Alzheimer's Disease
Catalog No. 50-210-8663