Learn More
TARGETMOL CHEMICALS INC BETA-AMYLOID 25-35 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T142845MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. (beta)-Amyloid (25-35) (A(beta)25-35) is the fragment A(beta)(25-35) of the Alzheimer's amyloid (beta)-peptide. Which has shown neurotoxic activities in cultured cells. purity: 97%
Catalog No. 50-258-1301