Learn More
TARGETMOL CHEMICALS INC beta-Amyrone 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN35001MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. beta-Amyrone (beta-Amyron) has antifungal activity against Chikungunya virus replication with an EC50 of 86 uM. beta-Amyrone has anti-inflammatory activity through inhibiting the expression of COX-2. beta-Amyrone exhibits anti-alpha-glucosidase inhibitory activity and moderate AChE activity. beta-Amyrone can be used in the research of disease like inflammation, infection, and obesity. Purity 99.55%
Catalog No. 50-288-0243