Learn More
Medchemexpress LLC beta-Catenin Mouse 50ug
SDP
Supplier:  Medchemexpress LLC HYP7438150UG
Encompass_Preferred
-Catenin is a key component of canonical Wnt signaling which dynamically controls cellular processes In the absence of Wnt it forms a complex leading to phosphorylation and degradation beta-Catenin Protein Mouse (sf9 His-GST) is the recombinant mouse-derived beta-Catenin protein expressed by Sf9 insect cells with N-His N-GST labeled tag
Catalog No. 50-001-78576