Learn More
Cusabio Technology LLC Human Beta-Endorphin,β-EP ELISA Kit
Supplier:  Cusabio Technology LLC CSBE06821H
Encompass_Preferred
This Human β-EP ELISA Kit was designed for the quantitative measurement of Human β-EP protein in serum, urine, cerebrospinal fluid (CSF), saliva, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 62.5 pg/mL-4000 pg/mL and the sensitivity is 15.6 pg/mL. One kit contains 96 wells.
Catalog No. NC2952876