Learn More
Medchemexpress LLC HY-N6018 5mg Medchemexpress, Beta-Eudesmol CAS:473-15-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN60185MG
Encompass_Preferred
Medchemexpress, HY-N6018 5mg Beta-Eudesmol CAS:473-15-4 Beta-Eudesmol is a natural oxygenated sesquiterpene, activates hTRPA1, with an EC50 of 32.5 μM. Beta-Eudesmol increases appetite through TRPA1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1556