Learn More
Sigma Aldrich Fine Chemicals Biosciences beta-Galactosidase from Es
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G60085KU
Encompass
-Galactosidase is a tetramer consisting of four equal subunits of 135 000 Da each. It is a sulfhydryl containing enzyme with 19 cysteine residues per subunit.
Catalog No. 50-290-2443