Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Galactosidase from Es
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G563515KU
Encompass
-Galactosidase is a tetramer consisting of four equal subunits of 135 000 each. It is a sulfhydryl containing enzyme with 19 cysteine residues per subunit.
Catalog No. 50-291-4466