Learn More
ABclonal Technology Beta-galactosidase GLB1
SDP
Supplier:  ABclonal Technology A23753100UL
Encompass_Preferred
This gene encodes a member of the glycosyl hydrolase 35 family of proteins Alternative splicing results in multiple transcript variants at least one of which encodes a preproprotein that is proteolytically processed to generate the mature lysosomal enzyme This enzyme catalyzes the hydrolysis of a terminal beta-linked galactose residue from ganglioside substrates and other glycoconjugates Mutations in this gene may result in GM1-gangliosidosis and Morquio B syndrome
Catalog No. 50-000-05511