Learn More
Medchemexpress LLC Beta-Glucocerebrosidase modulator 1 | 3080715-77-8 | 99.0% | 366.43 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY163779
Encompass_Preferred
β-Glucocerebrosidase modulator 1 (Compound 28) is an allosteric modulator for β-Glucocerebrosidase. It exhibits an EC50 of 9.0 μM and a Kd of 0.050 μM. This product is a light yellow to yellow oil and is intended for research use only.
  • Allosteric modulator for β-Glucocerebrosidase
  • EC50 of 9.0 μM
  • Kd of 0.050 μM
  • For research use only
  • Appearance: Light yellow to yellow oil
Catalog No. 50-003-65653