Learn More
Sigma Aldrich Fine Chemicals Biosciences beta-Glucuronidase from Escherichia coli Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay) | 9001-45-0 | MFCD00130629 | 25KU
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G739625KU
Encompass
beta-Glucuronidase from Escherichia coli Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay) | 9001-45-0 | MFCD00130629 | 25KU
Catalog No. 50-178-1237