Learn More
Medchemexpress LLC Beta-glucuronidase | 9001-45-0 | >95.0% | 100UG

Supplier: Medchemexpress LLC HYP70258100UG
Recombinant beta-glucuronidase (GUSB) expressed in Escherichia coli with an N-terminal 6xHis tag, supplied as a 0.2 μm filtered, stabilized solution for biochemical assays. The enzyme hydrolyzes glucuronide substrates (e.g., phenolphthalein glucuronide, PNPG, and 4-methylumbelliferyl-glucuronide) and is suitable for activity testing, biochemical studies, and assay development. Store frozen and avoid repeated freeze-thaw cycles.
- Recombinant enzyme expressed in E. coli for consistent activity.
- N-terminal His tag enables affinity purification and detection.
- Supplied as a filtered, stabilized solution for immediate use.
- High purity (>95%) suitable for sensitive assays.
- Active against common glucuronide substrates for activity testing.
- Stable at -80 °C for long-term storage; avoid repeated freeze-thaw.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.