Learn More
Medchemexpress LLC beta-Mangostin | 20931-37-7 | 424.49 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN094110MM/1ML
Encompass_Preferred
beta-Mangostin (β-Mangostin) is a xanthone compound found in *Cratoxylum arborescens*. It exhibits antibacterial and antimalarial activities, including antimycobacterial activity against *Mycobacterium tuberculosis* and *in vitro* antimalarial activity against *Plasmodium falciparum*. It also shows potent anticancer activity against various cancers, such as hepatocellular carcinoma and leukaemic.
  • Xanthone compound
  • Antibacterial activity
  • Antimalarial activity
  • Potent anticancer activity
Catalog No. 50-004-17030