Learn More
CPC Scientific H-Asp-Glu-Gly-Pro-Tyr-Lys-Met-Glu-His-Phe-Arg-Trp-Gly-Ser-Pro-Pro-Lys-Asp-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific MSH006B
Encompass
ONE-LETTER SEQUENCE: DEGPYKMEHFRWGSPPKDMOLECULAR FORMULA: C98H138N26O29S1MOLECULAR WEIGHT:2176.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [19941-13-0], SYNONYMS: melanotropinsRESEARCH AREA: Hormonal
Catalog No. 50-210-9832