Learn More
CPC Scientific H-Asp-Glu-Gly-Pro-Tyr-Arg-Met-Glu-His-Phe-Arg-Trp-Gly-Ser-Pro-Pro-Lys-Asp-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific MSH005B
Encompass
ONE-LETTER SEQUENCE: DEGPYRMEHFRWGSPPKDMOLECULAR FORMULA: C98H138N28O29S1MOLECULAR WEIGHT:2204.41STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [17750-75-3], SYNONYMS: melanotropinsRESEARCH AREA: Hormonal
Catalog No. 50-210-9830