Learn More
Medchemexpress LLC Beta-muricholic acid | 2393-59-1 | MFCD00271403 | 99.9% | 408.57 g·mol⁻¹ | C24H40O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1337075MG
Encompass_Preferred
β-Muricholic acid is a research-grade bile acid supplied as a high-purity solid for use as a biochemical standard and reagent in studies of bile acid metabolism, cholesterol homeostasis, and analytical assays. It is typically handled as a powder and requires protection from light with recommended cold storage for solutions.
  • High purity suitable for analytical and in vitro use.
  • Solid form for accurate weighing and dissolution.
  • Compatible with common solvents such as DMSO.
  • Stable when stored as recommended (protect from light, cold storage for solutions).
  • Suitable as a reference standard in bile acid and lipid metabolism studies.
Catalog No. 50-002-62692