Learn More
Medchemexpress LLC Beta-NGF Mouse 110 500ug

Supplier: Medchemexpress LLC HYP70530A500UG
Recombinant mouse beta-nerve growth factor (β-NGF) is a 110 amino-acid protein expressed in E. coli and supplied lyophilized with an N-terminal His tag. It is provided at >95% purity with a predicted mass of ~12.4 kDa and is intended for in vitro research on NGF/TrkA signaling, neuronal survival, and differentiation. Datasheet, COA, and handling instructions are available.
- Recombinant mouse NGF (110 a.a) with an N-terminal His tag for purification and detection.
- High purity greater than 95% as determined by reducing SDS-PAGE.
- Predicted molecular mass of 12.4 kDa and consistent SDS-PAGE profile.
- Lyophilized format expressed in E. coli for long-term stability.
- Multiple vial sizes available, including a 500 μg option for scale-up experiments.
- Recommended reconstitution concentration of at least 100 μg/mL and storage at -20°C or -80°C for extended storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.