Learn More
Medchemexpress LLC Beta-rubromycin | 27267-70-5 | 97.7% | 1 MG

Supplier: Medchemexpress LLC HY1224821MG
β-Rubromycin is a quinone-class natural product used as a research reagent with reported telomerase and HIV reverse transcriptase inhibitory activity. It is supplied as a solid for in vitro biochemical assays; consult the product data sheet and SDS for handling, storage, and safety information.
- Quinone antibiotic scaffold with telomerase and reverse transcriptase inhibition.
- High purity research material (97.7%).
- Formula C27H20O12; molecular weight 536.44 g/mol.
- Supplied as a 1 mg solid, soluble in DMSO for in vitro assays.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.