Learn More
CPC Scientific H-Leu-Pro-Phe-Phe-Asp-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AMYD032A
Encompass
Pentapeptide that inhibited amyloid formation in vitro and prevented amyloid neurotoxicity in cell culture. It also has the capacity to reduce in vivo cerebral Abeta deposition and to completely block amyloid fibril formation in rat brain.SEQUENCE: H-Leu-Pro-Phe-Phe-Asp-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: LPFFDMOLECULAR FORMULA: C33H43N5O8MOLECULAR WEIGHT: 637.73STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [182912-74-9]SYNONYMS: (Pro,Asp)-Amyloid -Protein (17-21)RESEARCH AREA: Alzheimer's DiseaseREFERENCES:Chacon, M. A., et al. "-sheet breaker peptide prevents A -induced spatial memory impairments with partial reduction of amyloid deposits." Molecular Psychiatry 9.10 (2004): 953-961SKU(s): AMYD-032A
Catalog No. 50-210-8690