Learn More
Medchemexpress LLC HY-N0171 100mg Medchemexpress, Beta-Sitosterol CAS:83-46-5 Purity:>80%
SDP
Supplier:  Medchemexpress LLC HYN0171100MG
Encompass_Preferred
Beta-Sitosterol (purity>80%) includes β-sitosterol (≥80%), stigmasterol, campesterol and brassicasterol mainly. Beta-Sitosterol is a plant sterol. Beta-Sitosterol (purity>80%) interfere with multiple cell signaling pathways, including cell cycle, apoptosis, proliferation, survival, invasion, angiogenesis, metastasis and inflammation.For research use only.
Catalog No. 50-193-3443