Learn More
Apexbio Technology LLC Betacarotene, 5g Cas# 7235-40-7
SDP
Supplier:  Apexbio Technology LLC A83775000
Encompass_Preferred
Beta -Carotene is a strongly colored red-orange pigment abundant in plants and fruits. 7235-40-7. MFCD00001556 -Carotene is a strongly colored red-orange pigment abundant in plants and fruits. It is an organic compound and chemically is classified as a hydrocarbon and specifically as a terpenoid, reflecting its derivation from isoprene units. -Carotene is biosynthesized from geranylgeranyl pyrophosphate. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8505