Learn More
Apexbio Technology LLC Betahistine 2HCl 5579-84-0 1g
SDP
Supplier:  Apexbio Technology LLC B15551000
Encompass_Preferred
Betahistine 2HCl (CAS 5579-84-0) is a small-molecule inhibitor targeting histamine H3 receptors exhibiting an inhibitory concentration (IC50) of 1 9 M By antagonizing H3 receptors betahistine modulates histaminergic neurotransmission which influences the release of various neurotransmitters in the central nervous system This compound is widely utilized in research focused on the physiological roles of histamine signaling particularly in studies of neurological and vestibular disorders as well as in the exploration of H3 receptor pharmacology and modulation
Catalog No. 50-000-87682