Learn More
Medchemexpress LLC Betahistine | 5638-76-6 | 99.8% | 1 ML

Supplier: Medchemexpress LLC HYB052410MM/1ML
Betahistine is an orally active histamine H1 receptor agonist and an H3 receptor antagonist, primarily used for the study of rheumatoid arthritis (RA).
- Molecular weight: 136.19
- Chemical formula: C8H12N2
- Appearance: Liquid, light yellow to yellow
- Solubility in DMSO: 100 mg/mL (requires ultrasonication)
- Recommended storage: 4°C (sealed, away from moisture); -80°C for up to 6 months or -20°C for up to 1 month in solvent.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.