Learn More
Medchemexpress LLC Betahistine-d3 (dihydrochloride) | 244094-72-2 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0524AS1MG
Encompass_Preferred
Betahistine-d3 (dihydrochloride) is the deuterium labeled Betahistine dihydrochloride, an orally active histamine H1 receptor agonist and H3 receptor antagonist. It is used for the study of rheumatoid arthritis (RA) and can serve as a tracer or internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled Betahistine dihydrochloride
  • Orally active histamine H1 receptor agonist and H3 receptor antagonist
  • Used in the study of rheumatoid arthritis (RA)
  • Suitable as a tracer for drug development
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08772