Learn More
Medchemexpress LLC Betahistine-d3 dihydrochloride | 244094-72-2 | 99.0% | 212.13 | C8H11D3Cl2N2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0524AS10MG
Encompass_Preferred
Betahistine-d3 dihydrochloride is a deuterium-labeled form of betahistine supplied for research and analytical applications as a stable isotope reference standard. It is intended for use as a tracer or internal standard in mass spectrometry and pharmacokinetic studies.
  • Deuterium-labeled betahistine for use as a stable isotope standard.
  • High purity suitable for analytical applications (reported purity 99.0%).
  • Supplied as the dihydrochloride salt with CAS 244094-72-2.
  • Molecular weight 212.13 and molecular formula C8H11D3Cl2N2.
  • Available in small research pack sizes suitable for trace analysis.
Catalog No. 50-002-97030