Learn More
Medchemexpress LLC Betahistine dihydrochloride | 5579-84-0 | 99.8% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB0524A
Encompass_Preferred
Betahistine dihydrochloride is an orally active histamine H1 receptor agonist and a H3 receptor antagonist. It is used for the study of rheumatoid arthritis (RA).
  • Orally active histamine H1 receptor agonist and H3 receptor antagonist
  • Used for the study of rheumatoid arthritis
  • Regulates cAMP formation in CHO cells
  • Ameliorates collagen-induced arthritis in mouse models
  • Has potential applications in various clinical conditions
Catalog No. 50-004-06533