Learn More
Medchemexpress LLC Betaine-13C3 | 309762-22-9 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0710S1MG
Encompass_Preferred
Betaine-13C3 (Trimethylglycine-13C3) is the 13C labeled isotope of Betaine. This natural compound is found ubiquitously in plants, animals, and microorganisms, and is an active methyl-donor that maintains normal DNA methylation patterns. It also acts as an osmolyte, helping to maintain cellular water and ion balance, particularly in heat-stressed birds, and may promote intestinal microbial fermentation activity.
  • Functions as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Maintains normal DNA methylation patterns.
  • Acts as an osmolyte to maintain cellular water and ion balance.
  • Improves avian capacity against heat stress.
  • Promotes various intestinal microbes against osmotic variations.
  • Improves microbial fermentation activity.
Catalog No. 50-004-05322