Learn More
Apexbio Technology LLC Betaine hydrochloride 590-46-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC N17005.1
Encompass_Preferred
Betaine hydrochloride (CAS 590-46-5) is a quaternary ammonium compound that serves as a methyl group donor in cellular metabolism notably participating in the methionine-homocysteine cycle to modulate homocysteine levels Extracted from natural sources such as wolfberry and Achyranthes this molecule is soluble in water and ethanol facilitating its use in various assay systems Betaine hydrochloride is frequently employed in biomedical research to investigate metabolic regulation methylation processes and osmoregulatory mechanisms The compound should be stored tightly sealed in cool dry conditions to maintain stability for experimental applications
Catalog No. 50-000-87751