Learn More
Medchemexpress LLC Betaine (Standard) | 107-43-7 | MFCD00012123 | 99.5% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0710R
Encompass_Preferred
Betaine, a natural compound found in various foods, acts as an active methyl-donor, maintaining normal DNA methylation. Ubiquitous in plants, animals, and microorganisms, it functions as an osmolyte, helping to maintain cellular water and ion balance and improving microbial fermentation activity. This product serves as an analytical standard and reference, commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Natural compound and active methyl-donor
  • Maintains normal DNA methylation patterns
  • Functions as an osmolyte, maintaining cellular water and ion balance
  • Improves microbial fermentation activity
  • Analytical standard for reference
  • Used in HPLC, GC, and MS research
Catalog No. 50-004-06493