Learn More
Medchemexpress LLC Betamethasone 21-valerate | 2240-28-0 | 98.0% | 476.58 | 100 MG

Supplier: Medchemexpress LLC HY139352100MG
Betamethasone 21-valerate is a corticosteroid ester with anti-inflammatory activities.
- Chemical formula: C27H37FO6
- Appearance: Solid
- Color: White to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.