Learn More
Medchemexpress LLC Betamethasone 21-valerate | 2240-28-0 | MFCD00271767 | 98.0% | C27H37FO6 | 10MG

Supplier: Medchemexpress LLC HY13935210MG
Betamethasone 21-valerate is a corticosteroid ester used as an analytical and research standard with anti-inflammatory activity. It is supplied as a white to off-white solid and is intended for analytical testing, formulation studies, and other laboratory research applications.
- Has anti-inflammatory corticosteroid activity.
- Supplied as a white to off-white solid.
- High purity (98.0%) suitable for analytical applications.
- Molecular formula C27H37FO6 and molecular weight 476.58 g/mol.
- Available in milligram-scale packages for research use.
- Intended for research and analytical purposes, not for therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.