Learn More
Medchemexpress LLC Betamethasone | 378-44-9 | MFCD00062969 | >=98.0% | C22H29FO5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13570R10MG
Encompass_Preferred
Betamethasone is a synthetic glucocorticoid corticosteroid with potent anti-inflammatory and immunosuppressive activity. Supplied as a high-purity analytical standard, it is intended for research and laboratory use including assay development, method validation, and as a reference material in chromatographic and spectrometric analyses.
  • High purity suitable for analytical use.
  • Verified identity and assay for reliable standardization.
  • Solid form for accurate weighing and easy dissolution.
  • Compatible with LC, GC, and MS analytical methods.
  • Applicable to assay development, method validation, and calibration.
Catalog No. 50-002-60122