Learn More
Apexbio Technology LLC Betamethasone Dipropionate 5593-20-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A83785.1
Encompass_Preferred
Betamethasone Dipropionate (CAS 5593-20-4) is a synthetic glucocorticoid receptor agonist targeting intracellular glucocorticoid receptors It is designed to activate these receptors thereby modulating inflammatory and immune response pathways Betamethasone Dipropionate exerts its biological activity primarily through enhancement of glucocorticoid receptor-mediated gene transcription leading to suppression of inflammatory mediator production and inhibition of inflammatory cell recruitment Based on these pharmacological properties Betamethasone Dipropionate holds research potential in investigations of glucocorticoid signaling inflammatory skin disorders (such as dermatosis) and topical drug formulation research aimed at optimizing local delivery while minimizing systemic effects
Catalog No. 50-000-96235