Learn More
Selleck Chemical LLC Betamethasone Dipropionate S1688-1g
SDP
Supplier:  Selleck Chemical LLC S16881G
Encompass_Preferred
Betamethasone Dipropionate (SCH 11460) is a glucocorticoid steroid with anti-inflammatory and immunosuppressive abilities
Catalog No. 50-311-1592