Learn More
Medchemexpress LLC Betamethasone valerate | 2152-44-5 | 99.8% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0727
Encompass_Preferred
Betamethasone valerate, also known as Betamethasone 17-valerate, is the 17-valerate ester of Betamethasone. It functions as a topical corticosteroid with anti-inflammatory activity and is used in the treatment of recurrent aphthous stomatitis. It inhibits the binding of radiolabeled glucocorticoid dexamethasone to human epidermis and mouse skin with IC50s of 5 and 6 nM, respectively.
  • Topical corticosteroid with anti-inflammatory properties
  • Used for recurrent aphthous stomatitis treatment
  • Inhibits radiolabeled glucocorticoid binding
  • For research use only
Catalog No. 50-004-50495