Learn More
Medchemexpress LLC Betamipron | 3440-28-6 | 98.7% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1127500MG
Encompass_Preferred
Betamipron is a chemical compound which is used together with Panipenem to inhibit Panipenem uptake into the renal tubule and prevent nephrotoxicity.
  • Used in conjunction with Panipenem
  • Inhibits Panipenem uptake into renal tubule
  • Prevents nephrotoxicity
  • Targets bacteria
  • Functions as an antibiotic
  • Part of anti-infection pathway
Catalog No. 50-004-13494