Learn More
Medchemexpress LLC Betaxolol (hydrochloride) | 63659-19-8 | 98.1% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0381100MG
Encompass_Preferred
Betaxolol hydrochloride is a selective beta1 adrenergic receptor blocker used in research for conditions such as hypertension and glaucoma. It functions by selectively blocking beta-1 adrenergic receptors, which primarily affect the heart and kidney, leading to reduced heart rate and blood pressure. Research suggests it may also attenuate anxiety-like behavior during cocaine withdrawal. It is noted to have less systemic beta2- and potentially beta1-adrenergic receptor blockade compared to other similar agents.
  • Selective beta1 adrenergic receptor blockade.
  • Applicable in hypertension and glaucoma research.
  • May be safer for patients with reactive airway disease than some other agents.
  • Can attenuate anxiety-like behavior in certain contexts.
Catalog No. 50-163-9380