Learn More
Medchemexpress LLC Betaxolol hydrochloride | 63659-19-8 | 98.12% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0381A100MG
Encompass_Preferred
Betaxolol Hydrochloride is a selective beta1 adrenergic receptor blocker that can be used for the research of hypertension and glaucoma. It is a cardioselective beta-adrenergic receptor blocking agent. It has been shown to diminish anxiety-like behavior during early withdrawal from chronic cocaine administration in rats, and it produces less systemic beta 2- and possibly beta 1-adrenergic receptor blockade than either timolol or levobunolol, potentially making it safer for patients with reactive airway disease.
  • Selective beta1 adrenergic receptor blocker
  • Used in research for hypertension and glaucoma
  • Cardioselective beta-adrenergic receptor blocking agent
  • Diminishes anxiety-like behavior during withdrawal from chronic cocaine administration in rats
  • Produces less systemic beta 2- and possibly beta 1-adrenergic receptor blockade
Catalog No. 50-004-07180