Learn More
Medchemexpress LLC HY-B1557 10mg Medchemexpress, Betazole CAS:105-20-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB155710MG
Encompass_Preferred
Medchemexpress, HY-B1557 10mg Betazole CAS:105-20-4 Betazole (Ametazole), a pyrazole analogue of histamine, is an orally active histamine H2 receptor agonist. Betazole induces gastric acid secretion and causes an immediate and significant increase in common bile duct pressure. Betazole is used as a diagnostic agent known as histalog for investigating gastric acid secretory capacity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2708