Learn More
Enterprise Technology Solutions Betco Corporation Carpet Cleaner, FiberPro, FASTDRAW 15, 2L, 4/CT, Green
SDP
Supplier:  Enterprise Technology Solutions BET4024700
Encompass_Preferred
Powerful blend of low-foaming detergents is specifically designed for steam or hot water extraction carpet cleaning by quickly cutting through soil and grease to brighten carpet fibers instantly. Use FiberPRO Es-Steam Carpet Cleaner in all types of carpet extractors. Special blend of surfactants rinses easily and leaves no residue that can lead to premature resoiling. It contains the patented deodorizing power of O3Z malodor counteractant to eliminate odors at their source. This highly concentrated carpet cleaner is not for use on stain-resistant carpets. As part of the FastDraw system, it is numbered (FastDraw 15) and color-coded to eliminate misuse. Spill-proof bottle helps prevent chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.
Catalog No. 50-007-90313