Learn More
Medchemexpress LLC BETd-246 25mg | 2140289-17-2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11556825MG
Encompass_Preferred
BETd-246 is a second-generation PROTAC-based BET bromodomain degrader that links cereblon and BET ligands to induce selective BET protein degradation. It is provided as a research-grade solid (light yellow) with reported purity of 98.34% and a molecular weight of 946.02, and is highly soluble in DMSO for in vitro and formulation use.
  • PROTAC mechanism for targeted BET protein degradation.
  • Reported selectivity and antitumor potency in preclinical studies.
  • High purity suitable for research applications.
  • High solubility in DMSO enabling concentrated stock solutions.
  • Recommended storage as solid at -20°C under inert atmosphere for stability.
Catalog No. 50-002-26638