Learn More
Selleck Chemical LLC Bethanechol chloride S2455-50mg
SDP
Supplier:  Selleck Chemical LLC S245550MG
Encompass_Preferred
Bethanechol chloride(Myocholine Urecholine) is a selective muscarinic receptor agonist without any effect on nicotinic receptors
Catalog No. 50-311-2694