Learn More
Apexbio Technology LLC Bethanechol chloride 590-63-6 10mM (in 1mL H2O)
SDP
Supplier:  Apexbio Technology LLC B15995.1
Encompass_Preferred
Bethanechol chloride (CAS 590-63-6) is a cholinergic parasympathomimetic agent acting specifically as a muscarinic receptor agonist Structurally classified as a choline carbamate bethanechol selectively activates muscarinic cholinergic receptors without influencing nicotinic receptor subtypes This receptor-specific stimulation leads to enhanced parasympathetic activity facilitating investigation into gastrointestinal motility urinary bladder function and cholinergic signaling pathways Bethanechol chloride is commonly employed in biomedical research involving cholinergic receptor pharmacology and autonomic nervous system physiology
Catalog No. 50-000-62469