Learn More
Apexbio Technology LLC Bethanechol chloride, 200mg Cas# 590-63-6
SDP
Supplier:  Apexbio Technology LLC B1599200
Encompass_Preferred
Bethanechol Chloride is a selective muscarinic receptor agonist without any effect on nicotinic receptors.Bethanechol is a parasympathomimetic choline carbamate that selectively stimulates muscarinic receptors without any effect on nicotinic receptors. Un Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8553