Learn More
Cayman Chemical BethncholchlorIde 25g
SDP
Supplier:  Cayman Chemical 2383025 G
Encompass_Preferred
A muscarinic acetylcholine receptor agonist (IC50s = 1,837, 25, 631, 317, and 393 µM for M1-5, respectively); inhibits M2-mediated increases in cyclic AMP induced by isoproterenol in guinea pig small intestine (IC50 = 127 µM); increases basal tone of isolated porcine intravesical ureter (EC50 = 4.27 µM); induces fluid secretion in the ileum, duodenum, and jejunum of anesthetized rats (60 UG/kg)
Catalog No. 50-299-4823